Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromo-2,6-difluorobenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

47999-1G 47999-25G 47999-500MG 47999-5G
76916-558EA 376.62 USD InStock
76916-558 76916-560 76916-562 76916-564
3,5-Dibromo-2,6-difluorobenzaldehyde ≥97%
3,5-Dibromo-2,6-difluorobenzaldehyde


Formula: C₇H₂Br₂F₂O
Storage Temperature: Refrigerator
CAS Number: 1803713-69-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty