Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOB CHEM USA
Warning

57565-1G 57565-25G 57565-500MG 57565-5G
76917-350EA 194.15 USD InStock
76917-350 76917-352 76917-354 76917-356
3-Bromo-4'-methyl-1,1'-biphenyl ≥97%
3-Bromo-4'-methyl-1,1'-biphenyl


Formula: C₁₃H₁₁Br
Storage Temperature: Refrigerator
MDL Number: MFCD6201388
CAS Number: 844856-52-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty