Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-chloro-3-methylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

15560-1G 15560-25G 15560-5G 15560-500MG
76896-080EA 99.73 USD InStock
76896-080 76896-082 76896-084 77644-682
2-Bromo-6-chloro-3-methylphenylboronic acid ≥97%
2-Bromo-6-chloro-3-methylphenylboronic acid


Formula: C₇H₇BBrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11111904
CAS Number: 1309980-97-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty