Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-hydroxy-4-methylbenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

32607-1G 32607-25G 32607-500MG 32607-5G
76930-986EA 260.99 USD InStock
76930-986 76930-988 76930-990 76930-992
5-Bromo-2-hydroxy-4-methylbenzaldehyde ≥97% research grade
5-Bromo-2-hydroxy-4-methylbenzaldehyde


Formula: C₈H₇BrO₂
MW: 215.044 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01123249
CAS Number: 311318-63-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty