Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-ethylbenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

30025-1G 30025-25G 30025-5G
76939-576EA 96.55 USD InStock
76939-576 76939-578 76939-580
Ethyl 4-ethylbenzoate ≥97% research grade
Ethyl 4-ethylbenzoate


Formula: C₁₁H₁₄O₂
MW: 178.228 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00238713
CAS Number: 36207-13-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty