Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetate ≥97% research grade

Supplier: AOB CHEM USA
Warning

57327-1G 57327-25G 57327-500MG 57327-5G
76943-374EA 183.54 USD InStock
76943-374 76943-376 76943-378 76943-380
tert-Butyl 2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetate ≥97% research grade
tert-Butyl 2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetate


Formula: C₁₁H₁₇BrN₂O₂
MW: 289.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30480471
CAS Number: 2013373-96-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty