Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(5-Methyl-2-(methylthio)phenyl)-1,3-dioxolane ≥97%

Supplier: AOB CHEM USA
Warning

108112-1G 108112-500MG 108112-5G 108112-25G
76882-076EA 464.68 USD InStock
76882-076 76882-078 76882-080 77657-182
2-(5-Methyl-2-(methylthio)phenyl)-1,3-dioxolane ≥97%
2-(5-Methyl-2-(methylthio)phenyl)-1,3-dioxolane
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD34821361
CAS Number: 1421631-09-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty