Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Cyclopentyloxy)pyrimidine-5-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

121743-1G 121743-25G 121743-500MG 121743-5G
76882-388EA 233.4 USD InStock
76882-388 76882-390 76882-392 76882-394
2-(Cyclopentyloxy)pyrimidine-5-carboxylic acid ≥97%
2-(Cyclopentyloxy)pyrimidine-5-carboxylic acid


Formula: C₁₀H₁₂N₂O₃
Storage Temperature: Ambient
CAS Number: 1072855-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty