Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

15020-1G 15020-25G 15020-5G
76882-928EA 62.6 USD InStock
76882-928 76882-930 76882-932
2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester ≥97% research grade
2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester


Formula: C₁₄H₂₂BN₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD07368246
CAS Number: 957198-30-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty