Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Piperidino-5-pyrimidineboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

15022-1G 15022-25G 15022-5G
76882-942EA 82.76 USD InStock
76882-942 76882-944 76882-946
2-Piperidino-5-pyrimidineboronic acid ≥97% research grade
2-Piperidino-5-pyrimidineboronic acid


Formula: C₉H₁₄BN₃O₂
MW: 207.0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375149
CAS Number: 1002128-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty