Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,6-trichlorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

15876-1G 15876-25G 15876-500MG 15876-5G
76883-234EA 409.51 USD InStock
76883-234 76883-236 76883-238 76883-240
2,3,6-trichlorophenylboronic acid ≥97%
2,3,6-trichlorophenylboronic acid


Formula: C₆H₄BCl₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18400154
CAS Number: 851756-53-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty