Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Difluoro-4-(methoxymethoxy)benzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

15989-1G 15989-25G 15989-5G 15989-500MG
76883-284EA 114.58 USD InStock
76883-284 76883-286 76883-288 77648-798
1,2-Difluoro-4-(methoxymethoxy)benzene ≥97% research grade
1,2-Difluoro-4-(methoxymethoxy)benzene


Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30179097
CAS Number: 749230-22-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty