Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Bromophenyl)-6-butyl-1,3,6,2-dioxazaborolane ≥95%

Supplier: AOB CHEM USA
Warning

16961-1G 16961-25G 16961-5G 16961-500MG
76883-504EA 62.6 USD InStock
76883-504 76883-506 76883-508 77648-930
2-(2-Bromophenyl)-6-butyl-1,3,6,2-dioxazaborolane ≥95%
2-(2-Bromophenyl)-6-butyl-1,3,6,2-dioxazaborolane


Formula: C₁₄H₂₁BBrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09750470
CAS Number: 1257641-07-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty