Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-(Pyrrolidin-1-yl)pyrimidin-5-yl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

18039-1G 18039-25G 18039-500MG 18039-5G
76883-888EA 210.06 USD InStock
76883-888 76883-890 76883-892 76883-894
(2-(Pyrrolidin-1-yl)pyrimidin-5-yl)boronic acid ≥97% research grade
(2-(Pyrrolidin-1-yl)pyrimidin-5-yl)boronic acid


Formula: C₈H₁₂BN₃O₂
MW: 193.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696938
CAS Number: 955374-13-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty