Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,3-Difluoro-4-iodophenyl)ethanone ≥97%

Supplier: AOB CHEM USA
Warning

18201-1G 18201-25G 18201-500MG 18201-5G
76883-946EA 171.87 USD InStock
76883-946 76883-948 76883-950 76883-952
1-(2,3-Difluoro-4-iodophenyl)ethanone ≥97%
1-(2,3-Difluoro-4-iodophenyl)ethanone


Formula: C₈H₅F₂IO
Storage Temperature: Refrigerator
MDL Number: MFCD28103449
CAS Number: 885590-86-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty