Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-6-methylbenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

18597-1G 18597-25G 18597-5G
76884-072EA 88.06 USD InStock
76884-072 76884-074 76884-076
2,3-Difluoro-6-methylbenzaldehyde ≥97%
2,3-Difluoro-6-methylbenzaldehyde


Formula: C₈H₆F₂O
Storage Temperature: Refrigerator
CAS Number: 949026-74-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty