Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-methylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

18697-1G 18697-25G 18697-500MG 18697-5G
76884-114EA 260.99 USD InStock
76884-114 76884-116 76884-118 76884-120
2,4-Dichloro-5-methylphenylboronic acid ≥97%
2,4-Dichloro-5-methylphenylboronic acid


Formula: C₇H₇BCl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD26045242
CAS Number: 1421934-04-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty