Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluorophenylboronic acid N-butyldiethanolamine ester ≥95% research grade

Supplier: AOB CHEM USA
Warning

19321-25G 19321-5G
76884-250EA 453.01 USD InStock
76884-250 76884-252
2,3-Difluorophenylboronic acid N-butyldiethanolamine ester ≥95% research grade
2,3-Difluorophenylboronic acid N-butyldiethanolamine ester


Formula: C₁₄H₂₀BF₂NO₂
MW: 283.122 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19105362
CAS Number: 1190989-00-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty