Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethyl-4-hydroxymethylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

19472-1G 19472-25G 19472-500MG 19472-5G
76884-286EA 231.28 USD InStock
76884-286 76884-288 76884-290 76884-292
2,3-Dimethyl-4-hydroxymethylphenylboronic acid ≥97%
2,3-Dimethyl-4-hydroxymethylphenylboronic acid


Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD16295269
CAS Number: 1451391-38-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty