Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Piperidin-1-ylmethyl)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

20749-1G 20749-25G 20749-500MG 20749-5G
76884-630EA 170.81 USD InStock
76884-630 76884-632 76884-634 76884-636
2-(Piperidin-1-ylmethyl)phenylboronic acid ≥97% research grade
2-(Piperidin-1-ylmethyl)phenylboronic acid


Formula: C₁₂H₁₈BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06801661
CAS Number: 878289-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty