Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoropyridine-4-carboxaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

21033-1G 21033-25G 21033-5G 21033-500MG
76884-684EA 101.85 USD InStock
76884-684 76884-686 76884-688 77649-252
2,3-Difluoropyridine-4-carboxaldehyde ≥97%
2,3-Difluoropyridine-4-carboxaldehyde


Formula: C₆H₃F₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD16610817
CAS Number: 1227502-65-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty