Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,5-Dichloro-4-pyridyl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

21555-1G 21555-25G 21555-500MG 21555-5G
76884-798EA 124.13 USD InStock
76884-798 76884-800 76884-802 76884-804
(3,5-Dichloro-4-pyridyl)methanol ≥97%
(3,5-Dichloro-4-pyridyl)methanol


Formula: C₆H₅Cl₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD06202840
CAS Number: 159783-46-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty