Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-6-fluorobenzamide ≥97%

Supplier: AOB CHEM USA
Warning

22112-1G 22112-25G 22112-500MG 22112-5G
76884-950EA 171.87 USD InStock
76884-950 76884-952 76884-954 76884-956
2,4-Dibromo-6-fluorobenzamide ≥97%
2,4-Dibromo-6-fluorobenzamide


Formula: C₇H₄Br₂FNO
Storage Temperature: Ambient
MDL Number: MFCD19105408
CAS Number: 904285-17-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty