Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-6-chlorophenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

22269-1G 22269-25G 22269-500MG 22269-5G
76884-974EA 260.99 USD InStock
76884-974 76884-976 76884-978 76884-980
2,3-Difluoro-6-chlorophenylboronic acid ≥97% research grade
2,3-Difluoro-6-chlorophenylboronic acid


Formula: C₆H₄BClF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12922584
CAS Number: 1310404-70-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty