Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-(Sec-butoxy)naphthalen-2-yl)boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

22497-1G 22497-25G 22497-500MG 22497-5G
76885-036EA 142.17 USD InStock
76885-036 76885-038 76885-040 76885-042
(6-(Sec-butoxy)naphthalen-2-yl)boronic acid ≥97%
(6-(Sec-butoxy)naphthalen-2-yl)boronic acid


Formula: C₁₄H₁₇BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16260727
CAS Number: 1333411-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty