Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Dichloro-2-iodo-3-methoxybenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

22692-1G 22692-25G 22692-500MG 22692-5G
76885-048EA 124.13 USD InStock
76885-048 76885-050 76885-052 76885-054
1,5-Dichloro-2-iodo-3-methoxybenzene ≥97% research grade
1,5-Dichloro-2-iodo-3-methoxybenzene


Formula: C₇H₅Cl₂IO
Storage Temperature: Ambient
MDL Number: MFCD21607156
CAS Number: 1271523-34-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty