Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-isobutoxybenzene ≥97%

Supplier: AOB CHEM USA
Warning

23091-1G 23091-25G 23091-500MG 23091-5G
76885-136EA 203.7 USD InStock
76885-136 76885-138 76885-140 76885-142
1-Bromo-4-isobutoxybenzene ≥97%
1-Bromo-4-isobutoxybenzene


Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD11127625
CAS Number: 30752-23-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty