Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[6-[4-(Trifluoromethyl)phenyl]-3-pyridyl]methanol ≥97% research grade

Supplier: AOB CHEM USA
Warning

23703-1G 23703-25G 23703-500MG 23703-5G
76885-292EA 251.44 USD InStock
76885-292 76885-294 76885-296 76885-298
[6-[4-(Trifluoromethyl)phenyl]-3-pyridyl]methanol ≥97% research grade
[6-[4-(Trifluoromethyl)phenyl]-3-pyridyl]methanol


Formula: C₁₃H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD06410448
CAS Number: 356058-13-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty