Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Difluoro-5-(methylsulfanyl)benzene ≥97%

Supplier: AOB CHEM USA
Warning

23914-1G 23914-25G 23914-500MG 23914-5G
76885-350EA 124.13 USD InStock
76885-350 76885-352 76885-354 76885-356
1,3-Difluoro-5-(methylsulfanyl)benzene ≥97%
1,3-Difluoro-5-(methylsulfanyl)benzene


Formula: C₇H₆F₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11617772
CAS Number: 54378-77-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty