Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Methoxy-5-methylphenylpyridin-3-yl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

24237-1G 24237-25G 24237-500MG 24237-5G
76885-406EA 439.22 USD InStock
76885-406 76885-408 76885-410 76885-412
(6-Methoxy-5-methylphenylpyridin-3-yl)methanol ≥97%
(6-Methoxy-5-methylphenylpyridin-3-yl)methanol


Formula: C₈H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12024294
CAS Number: 1355224-12-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty