Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Bromo-4-methylbenzyl)piperidine ≥97%

Supplier: AOB CHEM USA
Warning

24951-1G 24951-25G 24951-500MG 24951-5G
76885-568EA 290.69 USD InStock
76885-568 76885-570 76885-572 76885-574
1-(3-Bromo-4-methylbenzyl)piperidine ≥97%
1-(3-Bromo-4-methylbenzyl)piperidine


Formula: C₁₃H₁₈BrN
Storage Temperature: Ambient
MDL Number: MFCD22628241
CAS Number: 1414870-77-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty