Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-5-fluorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

25373-1G 25373-25G 25373-500MG 25373-5G
76885-654EA 290.69 USD InStock
76885-654 76885-656 76885-658 76885-660
2,4-Dibromo-5-fluorobenzoic acid ≥97%
2,4-Dibromo-5-fluorobenzoic acid


Formula: C₇H₃Br₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD23135587
CAS Number: 173410-26-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty