Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Chloro-2'-fluoro-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOB CHEM USA
Warning

25412-1G 25412-25G 25412-500MG 25412-5G
76885-692EA 233.4 USD InStock
76885-692 76885-694 76885-696 76885-698
4'-Chloro-2'-fluoro-2,2,2-trifluoroacetophenone ≥97%
4'-Chloro-2'-fluoro-2,2,2-trifluoroacetophenone


Formula: C₈H₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD02260841
CAS Number: 886370-99-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty