Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Difluoro-5-methylsulfonylbenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

25543-1G 25543-25G 25543-5G 25543-500MG
76885-724EA 117.76 USD InStock
76885-724 76885-726 76885-728 77649-546
1,3-Difluoro-5-methylsulfonylbenzene ≥97% research grade
1,3-Difluoro-5-methylsulfonylbenzene


Formula: C₇H₆F₂O₂S
MW: 192.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972174
CAS Number: 171421-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty