Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,5-Dibromo-4-methoxyphenyl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

25838-1G 25838-25G 25838-500MG 25838-5G
76885-776EA 260.99 USD InStock
76885-776 76885-778 76885-780 76885-782
(3,5-Dibromo-4-methoxyphenyl)methanol ≥97%
(3,5-Dibromo-4-methoxyphenyl)methanol


Formula: C₈H₈Br₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12047705
CAS Number: 114113-99-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty