Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,6-Dichloropyridin-3-yl)ethanol ≥97%

Supplier: AOB CHEM USA
Warning

25997-1G 25997-25G 25997-500MG 25997-5G
76885-850EA 290.69 USD InStock
76885-850 76885-852 76885-854 76885-856
1-(2,6-Dichloropyridin-3-yl)ethanol ≥97%
1-(2,6-Dichloropyridin-3-yl)ethanol


Formula: C₇H₇Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD23163273
CAS Number: 1375708-67-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty