Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluoro-2-methoxybenzyl alcohol ≥97%

Supplier: AOB CHEM USA
Warning

26042-1G 26042-25G 26042-500MG 26042-5G
76885-880EA 242.95 USD InStock
76885-880 76885-882 76885-884 76885-886
3,4-Difluoro-2-methoxybenzyl alcohol ≥97%
3,4-Difluoro-2-methoxybenzyl alcohol


Formula: C₈H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28519671
CAS Number: 1779895-68-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty