Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromo-2-isopropylphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOB CHEM USA
Warning

27091-1G 27091-25G 27091-500MG 27091-5G
76886-134EA 377.69 USD InStock
76886-134 76886-136 76886-138 76886-140
(4-Bromo-2-isopropylphenyl)(thiomorpholino)methanone ≥97%
(4-Bromo-2-isopropylphenyl)(thiomorpholino)methanone


Formula: C₁₄H₁₈BrNOS
Storage Temperature: Refrigerator
MDL Number: MFCD32631797
CAS Number: 2364585-28-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty