Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4,5-difluoro-3-ethoxybenzene ≥97%

Supplier: AOB CHEM USA
Warning

27157-1G 27157-25G 27157-5G 27157-500MG
76886-160EA 96.55 USD InStock
76886-160 76886-162 76886-164 77649-702
1-Bromo-4,5-difluoro-3-ethoxybenzene ≥97%
1-Bromo-4,5-difluoro-3-ethoxybenzene


Formula: C₈H₇BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11036051
CAS Number: 204654-92-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty