Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Fluoro-4,5-dimethoxyphenyl)ethan-1-one 95%

Supplier: AOB CHEM USA
Warning

27873-1G 27873-25G 27873-5G 27873-500MG
76886-274EA 96.55 USD InStock
76886-274 76886-276 76886-278 77645-670
1-(2-Fluoro-4,5-dimethoxyphenyl)ethan-1-one 95%
1-(2-Fluoro-4,5-dimethoxyphenyl)ethan-1-one


Formula: C₁₀H₁₁FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD17226285
CAS Number: 142265-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty