Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Benzenedicarboxylic acid, 5-bromo-2-iodo-, 1,3-dimethyl ester ≥97%

Supplier: AOB CHEM USA
Warning

28315-25G 28315-5G
76886-406EA 595.17 USD InStock
76886-406 76886-408
1,3-Benzenedicarboxylic acid, 5-bromo-2-iodo-, 1,3-dimethyl ester ≥97%
1,3-Benzenedicarboxylic acid, 5-bromo-2-iodo-, 1,3-dimethyl ester


Formula: C₁₀H₈BrIO₄
Storage Temperature: Ambient
MDL Number: MFCD28360684
CAS Number: 1428236-23-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty