Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromothiazol-2-yl)ethanone ≥97%

Supplier: AOB CHEM USA
Warning

28805-1G 28805-25G 28805-500MG 28805-5G
76886-564EA 221.73 USD InStock
76886-564 76886-566 76886-568 76886-570
1-(4-Bromothiazol-2-yl)ethanone ≥97%
1-(4-Bromothiazol-2-yl)ethanone


Formula: C₅H₄BrNOS
Storage Temperature: Refrigerator
MDL Number: MFCD10697512
CAS Number: 208264-53-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty