Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Trityl-1H-pyrazol-4-yl)boronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

29421-1G 29421-500MG 29421-5G 29421-25G
76886-726EA 472.11 USD InStock
76886-726 76886-728 76886-730 77658-518
(1-Trityl-1H-pyrazol-4-yl)boronic acid ≥95% research grade
(1-Trityl-1H-pyrazol-4-yl)boronic acid


Formula: C₂₂H₁₉BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD22571854
CAS Number: 207307-51-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty