Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,3-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOB CHEM USA
Warning

30988-25G 30988-5G
76887-042EA 283.27 USD InStock
76887-042 76887-044
2-(2,3-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(2,3-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane


Formula: C₁₄H₂₁BO₄
MW: 264.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923194
CAS Number: 488850-92-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty