Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-methyl 3-(2,3-difluorophenyl)acrylate ≥97% research grade

Supplier: AOB CHEM USA
Warning

32128-1G 32128-25G 32128-5G
76887-402EA 62.6 USD InStock
76887-402 76887-404 76887-406
(E)-methyl 3-(2,3-difluorophenyl)acrylate ≥97% research grade
(E)-methyl 3-(2,3-difluorophenyl)acrylate


Formula: C₁₀H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD24134896
CAS Number: 1363402-34-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty