Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Propan-2-yloxy)pyridine-4-carboxylic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

33240-1G 33240-25G 33240-500MG 33240-5G
76887-564EA 201.58 USD InStock
76887-564 76887-566 76887-568 76887-570
(2-Propan-2-yloxy)pyridine-4-carboxylic acid ≥95% research grade
(2-Propan-2-yloxy)pyridine-4-carboxylic acid


Formula: C₉H₁₁NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09054678
CAS Number: 862507-33-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty