Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Isopropylphenyl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

33702-1G 33702-25G 33702-500MG 33702-5G
76887-664EA 350.1 USD InStock
76887-664 76887-666 76887-668 76887-670
(3-Isopropylphenyl)methanol ≥97%
(3-Isopropylphenyl)methanol


Formula: C₁₀H₁₄O
Storage Temperature: Refrigerator
MDL Number: MFCD07780489
CAS Number: 51473-70-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty