Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-(Benzyloxy)-2-methylphenyl)methanol ≥97% research grade

Supplier: AOB CHEM USA
Warning

34519-1G 34519-25G 34519-500MG 34519-5G
76887-864EA 257.8 USD InStock
76887-864 76887-866 76887-868 76887-870
(5-(Benzyloxy)-2-methylphenyl)methanol ≥97% research grade
(5-(Benzyloxy)-2-methylphenyl)methanol


Formula: C₁₅H₁₆O₂
MW: 228.286 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619498
CAS Number: 1450931-37-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty