Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-((4-(Tert-butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

35436-1G 35436-25G 35436-500MG 35436-5G
76888-038EA 223.85 USD InStock
76888-038 76888-040 76888-042 76888-044
(3-((4-(Tert-butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid ≥95% research grade
(3-((4-(Tert-butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid


Formula: C₁₆H₂₅BN₂O₄
MW: 320.192 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22419048
CAS Number: 865314-28-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty