Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyloxybenzyl alcohol ≥95%

Supplier: AOB CHEM USA
Warning

36124-25G 36124-5G
76888-210EA 177.18 USD InStock
76888-210 76888-212
3-Benzyloxybenzyl alcohol ≥95%
3-Benzyloxybenzyl alcohol
Precaución: for Research Use Only


Formula: C₁₄H₁₄O₂
MW: 214.26
Boiling Pt: 160
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00004642
CAS Number: 1700-30-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty